Learn More
Medchemexpress LLC HY-18341B 1g , L-Thyroxine (sodium) CAS:55-03-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY18341B1G
Encompass_Preferred
HY-18341B 1g L-Thyroxine (sodium) CAS: 55-03-8 L-Thyroxine sodium (Levothyroxine sodium) is a synthetic hormone for the research of hypothyroidism. DIO enzymes convert biologically active thyroid hormone (Triiodothyronine,T3) from L-Thyroxine (T4). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1017