Learn More
Medchemexpress LLC L-Thyroxine (Standard) | 51-48-9 | 98.7% | 776.87 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY18341R100MG
Encompass_Preferred
L-Thyroxine (Standard) is primarily used as an analytical standard for L-Thyroxine, making it suitable for research and analytical applications. It serves as a reference standard in various qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS. This synthetic hormone is also utilized in the study of hypothyroidism, where DIO enzymes are known to convert it into the biologically active thyroid hormone, Triiodothyronine (T3).
  • Analytical standard for L-Thyroxine.
  • Intended for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Commonly applied in techniques like HPLC, GC, and MS.
  • Synthetic hormone for hypothyroidism research.
  • Converted to triiodothyronine (T3) by DIO enzymes.
Catalog No. 50-003-87613