Learn More
Medchemexpress LLC L-Thyroxine (Standard) | 51-48-9 | 98.7% | 776.87 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY18341R50MG
Encompass_Preferred
L-Thyroxine (Standard) is the analytical standard of L-Thyroxine, intended for research and analytical applications. L-Thyroxine (Levothyroxine; T4) is a synthetic hormone used in the research of hypothyroidism. DIO enzymes convert biologically active thyroid hormone (Triiodothyronine, T3) from L-Thyroxine (T4).
  • Analytical standard for L-Thyroxine.
  • Intended for research and analytical applications.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-003-87664