Learn More
Medchemexpress LLC L-Triguluronic acid | 66754-14-1 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN7701A1MG
Encompass_Preferred
L-Triguluronic acid is a linear polysaccharide copolymer made of three L-guluronic acid (G) units, used in the formation of Alginate. Alginate, an unbranched polyanionic polysaccharide, is valuable for researching anti-fungal agent delivery carriers. Its properties, including non-toxicity, biodegradability, biocompatibility, mucoadhesive nature, and non-immunogenicity, make it suitable for drug delivery systems, particularly in treating fungal infections.
  • Composed of three L-guluronic acid (G) units
  • Can be used to form alginate
  • Alginate is an unbranched polyanionic polysaccharide
  • Useful for research into anti-fungal agent delivery carriers
  • Alginate exhibits non-toxicity, biodegradability, and biocompatibility
  • Mucoadhesive and non-immunogenic properties
  • Alginate-based systems show potential in treating fungal infections
Catalog No. 50-004-19635