Learn More
Medchemexpress LLC L-Tryptophan-1-13C | 81201-92-5 | 205.22 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0623S31MG
Encompass_Preferred
L-Tryptophan-1-13C is the 13C-labeled L-Tryptophan. L-Tryptophan (Tryptophan) is an essential amino acid that is the precursor of serotonin, melatonin, and vitamin B3. This product is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-88596