Learn More
Medchemexpress LLC L-Tryptophan-13C | 115709-68-7 | 205.22 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0623S15MG
Encompass_Preferred
L-Tryptophan-13C is the 13C-labeled L-Tryptophan, an essential amino acid that is a precursor of serotonin, melatonin, and vitamin B3.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-88780