Learn More
Medchemexpress LLC L-tryptophan-13C | 115709-68-7 | 99.9% | 205.22 g·mol⁻¹ | C10(13C)H12N2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0623S110MG
Encompass_Preferred
L-Tryptophan-13C is the 13C-labeled form of L-tryptophan used as a tracer and internal standard in analytical chemistry. It is suitable for quantitative analysis by NMR, GC-MS, and LC-MS, and is supplied in mg-scale quantities for research applications.
  • Isotopically labeled 13C L-tryptophan for tracer studies.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • High purity (reported at 99.93%).
  • Solid, white to off-white physical form.
  • Available in small research-scale packages (for example, 10 mg).
Catalog No. 50-002-96912