Learn More
Medchemexpress LLC L-Tryptophan-13C11 | 202114-65-6 | 215.14 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0623S21MG
Encompass_Preferred
L-Tryptophan-13C11 is the 13C-labeled L-Tryptophan. L-Tryptophan is an essential amino acid that acts as a precursor for serotonin, melatonin, and vitamin B3. This compound is suitable for use as a tracer and as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules
  • Used as a tracer for quantitation during the drug development process
  • Offers the potential to affect the pharmacokinetic and metabolic profiles of drugs
  • Purity: 99.78%
Catalog No. 50-003-88575