Learn More
Medchemexpress LLC L-tryptophan-15N | 205.22 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0623S75MG
Encompass_Preferred
L-Tryptophan-15N is the 15N-labeled L-Tryptophan. L-Tryptophan (Tryptophan) is an essential amino acid that is the precursor of serotonin, melatonin, and vitamin B3.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-88369