Learn More
Medchemexpress LLC L-tryptophan-15N2 | 204634-20-8 | MFCD00209799 | 99.9% | 206.21 | C11H12(15N)2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0623S810MG
Encompass_Preferred
L-Tryptophan-15N2 is the nitrogen-15 doubly labeled form of the essential amino acid L-tryptophan, supplied as a white to off-white solid for use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS analyses and metabolic flux studies.
  • Isotope-labeled (15N2) for tracer and internal standard applications.
  • High purity - 99.9% suitable for analytical workflows.
  • Supplied as a stable solid for convenient storage and handling.
  • Recommended storage: 4°C protected from light; in solution: -80°C (6 months), -20°C (1 month).
  • Compatible with NMR, GC-MS, and LC-MS quantitative assays.
Catalog No. 50-002-97995