Learn More
Medchemexpress LLC L-Tryptophan-α,β,β,2,4,5,6,7-d8 | 1233395-93-1 | 212.27 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0623S550MG
Encompass_Preferred
L-Tryptophan-d8 is the deuterium labeled L-Tryptophan. L-Tryptophan is an essential amino acid that is the precursor of serotonin, melatonin, and vitamin B3. This compound is suitable for use as a tracer or an internal standard in quantitative analysis techniques.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-88703