Learn More
Medchemexpress LLC L-Tryptophan | 73-22-3 | 204.23 | 100 G
SDP
Supplier:  Medchemexpress LLC HYN0623100G
Encompass_Preferred
L-Tryptophan is an orally active and essential amino acid that is the precursor of serotonin, melatonin, and vitamin B3. It can promote an increase in stemness and osteogenic ability of BMSCs in vitro and in vivo, and inhibits cell proliferation and induces cell cycle arrest with high levels.
  • Orally active and essential amino acid
  • Precursor of serotonin, melatonin, and vitamin B3
  • Promotes an increase in stemness and osteogenic ability of BMSCs in vitro and in vivo
  • Inhibits cell proliferation and induces cell cycle arrest with high levels
Catalog No. 50-003-88634