Learn More
Medchemexpress LLC L-tryptophan, L-valyl- | 24587-37-9 | 97.9% | 303.36 g/mol | C16H21N3O3 | 1 ML

Supplier: Medchemexpress LLC HY11806010MM 1ML
Dipeptide 2 (N-Valyltryptophan) is a bioactive dipeptide reported to have anti-aging activity and used as a cosmetic ingredient and research reagent. It is supplied as a white to off-white solid or as a ready-to-use 10 mM solution in DMSO, and requires low-temperature storage to maintain stability.
- CAS No. 24587-37-9
- Purity 97.9%
- Molecular formula C16H21N3O3; MW 303.36 g/mol
- Solubility: DMSO 12.5 mg/mL (41.2 mM); ultrasonic required
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months)
- Available as 10 mM, 1 mL solution in DMSO or in solid pack sizes
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.