Learn More
Cayman Chemical L-Tryptophn-d5 25mg
SDP
Supplier:  Cayman Chemical 3482925 MG
Encompass_Preferred
An internal standard for the quantification of L-tryptophan by GC- or LC-MS
Catalog No. 50-291-0191