Learn More
Cayman Chemical AflatoxIn B1-13C17 1.2mL
SDP
Supplier:  Cayman Chemical 312601.2 ML
Encompass_Preferred
An internal standard for the quantification of aflatoxin B1 by GC- or LC-MS
Catalog No. 50-299-1471