Learn More
Medchemexpress LLC L-Tyrosinamide, L-tyrosyl-L-asparaginyl-L-tryptophyl-L-asparaginyl-L-seryl-L-phenylalanylglycyl-L-leucyl-L-arg | 478507-53-8 | 99.6% | 1432.46 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP1197A
Encompass_Preferred
Kisspeptin-10 (mouse, rat) TFA is a potent vasoconstrictor and inhibitor of angiogenesis. It acts as a ligand for the rodent kisspeptin receptor (KISS1, GPR54) and has been shown to reduce Methotrexate-induced reproductive toxicity, acting as a potential antioxidant compound.
  • Potent vasoconstrictor
  • Inhibits angiogenesis
  • Ligand for the rodent kisspeptin receptor (KISS1, GPR54)
  • Reduces methotrexate-induced reproductive toxicity
  • Acts as a potential antioxidant compound
Catalog No. 50-004-26996