Learn More
Medchemexpress LLC L-Tyrosine-13C9 | 55443-60-2 | 190.12 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0473S35MG
Encompass_Preferred
L-Tyrosine-13C9 is a 13C-labeled L-Tyrosine, a non-essential amino acid that can inhibit citrate synthase activity in the posterior cortex. It is suitable for research applications.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-89357