Learn More
Medchemexpress LLC L-Tyrosine-15N | 35424-81-8 | 182.18 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0473S1100MG
Encompass_Preferred
L-Tyrosine-15N is the 15N-labeled L-Tyrosine, a non-essential amino acid that can inhibit citrate synthase activity in the posterior cortex. It can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have gained attention for their potential to affect the pharmacokinetic and metabolic profiles of drugs, making this compound valuable for research applications.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Useful in metabolic flux analysis
  • Suitable for SILAC Reagents
  • Applicable in newborn screening
Catalog No. 50-003-88946