Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Tyrosine-3,5-13C2 | 70479-98-0 | 183.17 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN0473S65MG

Encompass_Preferred

L-Tyrosine-3,5-13C2 is the 13C-labeled L-Tyrosine. This non-essential amino acid can inhibit citrate synthase activity in the posterior cortex. Stable heavy isotopes have been incorporated into drug molecules for use as tracers in quantitation during drug development. It can also be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.

  • For research use only
  • Purity: 99.43%
  • Appearance: Solid
  • Color: White to off-white

Catalog No. 50-003-89239


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.