Learn More
Medchemexpress LLC L-Tyrosine, 3-hydroxy-, methyl ester, hydrochloride (1:1) | 1421-65-4 | 99.8% | 247.68 | 1 G

Supplier: Medchemexpress LLC HYW0119631G
Melevodopa hydrochloride is an effervescent Levodopa prodrug. Levodopa is an orally active metabolic precursor of neurotransmitters dopamine and has anti-allodynic effects and the potential for Parkinson's disease.
- Solid appearance
- White to off-white color
- Soluble in DMSO
- Store at 4°C under nitrogen
- Store in solvent at -80°C for 6 months, or -20°C for 1 month under nitrogen
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.