Learn More
Medchemexpress LLC L-Tyrosine-d2-1 | 30811-19-9 | 183.20 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYN0473S10
Encompass_Preferred
L-Tyrosine-d2-1 is a deuterium labeled version of L-Tyrosine, a non-essential amino acid known to inhibit citrate synthase activity in the posterior cortex. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution can influence pharmacokinetic and metabolic profiles.
Catalog No. 50-003-89452