Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-tyrosine-d2-2 | 57746-15-3 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN0473S1310MG

Encompass_Preferred

Deuterium-labeled L-tyrosine used as a stable-isotope standard and tracer for biochemical, pharmacokinetic, and mass spectrometry applications.

  • Deuterium-labeled amino acid for tracer and quantitation studies.
  • Molecular formula C9H9D2NO3; molecular weight 183.20 g/mol.
  • CAS number 57746-15-3.
  • Suitable as an internal standard in LC-MS/MS workflows.
  • Storage: powder at -20°C; in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Typical pack size: 10 MG.

Catalog No. 50-002-95767


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.