Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Tyrosine-β,β-d2 | 72963-27-0 | 183.20 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYN0473S8

Encompass_Preferred

L-Tyrosine-d2 is a deuterium-labeled version of L-Tyrosine, a non-essential amino acid known to inhibit citrate synthase activity in the posterior cortex. This stable isotope is intended for research purposes.

  • Deuterium labeled L-Tyrosine.
  • Purity: 99.81%.
  • Appearance: Solid, white to off-white.
  • Molecular Weight: 183.20.
  • Formula: C9H9D2NO3.
  • CAS No.: 72963-27-0.
  • Storage: 4°C, sealed storage, away from moisture and light. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture and light).
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Intended for research use only.

Catalog No. 50-003-89814


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.