Learn More
Alkali Scientific L Tyrosine, Disodium Salt Dihydrate, 1 Kg
SDP
Supplier:  Alkali Scientific LTY003CP
Encompass_Preferred
L-Tyrosine (1kg) is an amino acid that is a precursor to important neurotransmitters involved in regulating mood, stress, and cognitive performance. This amino acid is commonly used in research related to mental clarity, cognitive function, and the effects of stress. L-Tyrosine is involved in the synthesis of dopamine, norepinephrine, and epinephrine, all of which are critical for brain health and function. The 1kg bottle is ideal for large-scale laboratory research, clinical studies, and as an ingredient in dietary supplements focused on improving mood, focus, and energy levels. Tyrosine supplementation has shown potential benefits in managing stress and improving performance under pressure.
Catalog No. 50-006-60086