Learn More
Medchemexpress LLC L-tyrosine hydrazide | 7662-51-3 | MFCD00007609 | 99.6% | 195.22 g/mol | C9H13N3O2 | 5 G

Supplier: Medchemexpress LLC HYW1420005G
L-Tyrosine hydrazide is a tyrosine-derived amino acid reagent provided for research use. It is a hydrazide derivative of L-tyrosine commonly used as a synthetic intermediate and biochemical reagent in studies of amino acid metabolism, derivatization chemistry, and analytical method development. Supplied as a high-purity solid suitable for analytical and preparative applications.
- High purity: 99.6%.
- Molecular weight: 195.22 g/mol.
- Chemical formula: C9H13N3O2.
- Appearance: white to off-white solid.
- Solubility: soluble in 1M HCl (50 mg/mL) and in DMSO (5 mg/mL).
- Storage: store protected from light; 4°C recommended; in solution store at -80°C for long term.
- Pack size: 5 G.
- Intended use: for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.