Learn More
Medchemexpress LLC L-tyrosine, N-[(1,1-dimethylethoxy)carbonyl]-, 1,1-dimethylethyl ester | 18938-60-8 | MFCD20264826 | 99.8% | 337.41 g/mol | C18H27NO5 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW019676500G
Encompass_Preferred
Boc-Tyr-OtBu is the N-Boc, tert-butyl ester protected derivative of L-tyrosine, supplied as a white to off-white solid. It serves as a protected amino acid building block for peptide and small-molecule synthesis, enabling selective deprotection and coupling strategies.
  • Provides N-Boc protection of the amino group
  • Includes tert-butyl ester protection of the carboxyl group for stability during coupling reactions
  • High purity (~99.8%) for consistent reaction performance
  • Soluble in common organic solvents used in peptide chemistry
  • Suitable for solid-phase and solution-phase peptide synthesis
Catalog No. 50-003-00094