Learn More
Medchemexpress LLC L-Tyrosine, O-methyl- | 6230-11-1 | 99.74% | 195.22 | 10 G

Supplier: Medchemexpress LLC HYW00991210G
H-Tyr(Me)-OH is a synthetic amino acid that can enter into protein in *E. coli* in response to an amber nonsense codon. It is intended for research use only.
- Synthetic amino acid product type
- Intended for research use only
- Appears as a solid
- Color is white to off-white
- Powder storage: -20°C for 3 years; 4°C for 2 years
- Storage in solvent: -80°C for 6 months; -20°C for 1 month
- Soluble in H2O at 2 mg/mL (10.24 mM; requires ultrasonic)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.