Learn More
Medchemexpress LLC L-Valine-13C5 | 55443-52-2 | 122.11 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0717S65MG
Encompass_Preferred
L-Valine-13C5 ((S)-Valine-13C5) is the 13C-labeled version of L-Valine. L-Valine is a nonlinear semiorganic material that causes lipid peroxidation and accumulation of malondialdehyde (MDA), and it exhibits inhibitory activity against cyanobacteria. It also inhibits multidrug-resistant bacteria by activating the PI3K/Akt signaling pathway and inhibiting arginase. This compound is intended for research use only and can be used as a tracer or as an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS
  • Exhibits inhibitory activity against cyanobacteria
  • Inhibits multidrug-resistant bacteria by activating the PI3K/Akt signaling pathway and inhibiting arginase
Catalog No. 50-004-16751