Learn More
Medchemexpress LLC L-Valine-15N | 59935-29-4 | 118.14 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0717S100MG
Encompass_Preferred
L-Valine-15N is a 15N-labeled version of L-Valine, a nonlinear semiorganic material. It causes lipid peroxidation and accumulation of malondialdehyde, and exhibits inhibitory activity against cyanobacteria. This compound also inhibits multidrug-resistant bacteria by activating the PI3K/Akt signaling pathway and inhibiting arginase.
  • Intended for research use only.
  • Can be utilized as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-15303