Learn More
Medchemexpress LLC L-Valine-15N | 59935-29-4 | 118.14 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0717S
Encompass_Preferred
L-Valine-15N is the 15N-labeled L-Valine, a nonlinear semiorganic material. It causes lipid peroxidation and accumulation of malondialdehyde (MDA), exhibits inhibitory activity against cyanobacteria, and inhibits multidrug-resistant bacteria through activation of PI3K/Akt signaling pathway and inhibition of arginase. This product is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-90216