Learn More
Medchemexpress LLC L-valine, N-[(phenylmethoxy)carbonyl]-L-alanyl- | 14550-79-9 | MFCD00026455 | 96.5% | 322.36 g/mol | C16H22N2O5 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0488281G
Encompass_Preferred
((Benzyloxy)carbonyl)-L-alanyl-L-valine is a carbobenzoxy-protected dipeptide (benzyloxycarbonyl L-alanyl-L-valine) supplied for research use. It is a valine derivative used in peptide chemistry and biochemical assays. Typical physical data include CAS 14550-79-9, formula C16H22N2O5, molecular weight 322.36 g/mol, and manufacturer-reported purity ~96.5%. Storage: powder at -20°C (3 years) or 4°C (2 years); in solution at -80°C (6 months) or -20°C (1 month).
  • Carbobenzoxy-protected L-alanyl-L-valine dipeptide for research use
  • Valine derivative commonly used in peptide synthesis and biochemical studies
  • Manufacturer-reported purity near 96.5% for analytical and preparative applications
  • Available in small research sizes; store as recommended to preserve stability
  • Provided as a powder suitable for dissolution in common organic solvents
Catalog No. 50-002-10569