Learn More
Medchemexpress LLC L-valyl-L-phenylalanine | 3918-92-1 | MFCD00020423 | 99.1% | 264.32 g·mol⁻1 | C14H20N2O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10737810MM 1ML
Encompass_Preferred
L-valyl-L-phenylalanine is a dipeptide (Val-Phe) supplied as a ready-to-use 10 mM solution in water (1 mL) for research and analytical applications. It is used as an endogenous metabolite standard and in biochemical assays where a high-purity dipeptide reagent is required.
  • High purity (99.1%) for reliable analytical results.
  • Supplied as a 10 mM solution to enable immediate use in assays.
  • Compact 1 mL format for small-volume experiments.
  • Stable storage recommendations provided for powder and solution forms.
  • Suitable as an analytical standard and for metabolite research.
Catalog No. 50-002-41258