Learn More
Medchemexpress LLC L-valyl-L-tryptophan | 24587-37-9 | 97.9% | 303.36 g/mol | C16H21N3O3 | 1 G

Supplier: Medchemexpress LLC HY1180601G
Dipeptide 2 (N-Valyltryptophan) is a small bioactive dipeptide reported to have anti-aging activity and commonly used as a cosmetic ingredient and in skin biology research. It is supplied as a white to off-white solid and is available in multiple packaging sizes and as a DMSO solution.
- Small bioactive dipeptide with reported anti-aging effects.
- High purity (97.9%) suitable for research applications.
- Available as solid powder and as a 10 mM solution in DMSO.
- Stable when stored as powder at -20°C or refrigerated at 4°C for shorter terms.
- Molecular weight 303.36 g/mol; formula C16H21N3O3; CAS 24587-37-9.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.