Learn More
Medchemexpress LLC L-Valyl-N-(4-(hydroxymethyl)phenyl)-L-alaninamide | 1343476-44-7 | MFCD32641649 | 99.7% | 293.36 g/mol | C15H23N3O3 | 250MG

Supplier: Medchemexpress LLC HY125933250MG
Val-Ala-PAB is a cleavable antibody-drug conjugate (ADC) linker composed of a valine-alanine dipeptide and a para-aminobenzyl (PAB) self-immolative spacer, designed to enable enzymatic release of payloads in target cells. Provided as a high-purity research reagent, it is intended for linker development and conjugation chemistry in ADC synthesis.
- Cleavable linker that enables enzymatic release of payloads.
- Contains a valine-alanine dipeptide and PAB self-immolative spacer.
- High purity suitable for research applications.
- Molecular weight approximately 293.36 g/mol.
- Chemical formula C15H23N3O3.
- Soluble in DMSO; ultrasonic assistance may improve dissolution.
- Store protected from light; follow refrigerated or frozen conditions for solutions.
- Available in small research package sizes, including 250 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.