Learn More
Medchemexpress LLC L-Valyl-N-(4-(hydroxymethyl)phenyl)-L-alaninamide | 1343476-44-7 | MFCD32641649 | 99.7% | 293.37 g/mol | C15H23N3O3 | 1G
SDP
Supplier:  Medchemexpress LLC HY1259331G
Encompass_Preferred
Val-Ala-PAB is a peptide-based, enzymatically cleavable linker designed for antibody-drug conjugate (ADC) synthesis. It couples a valine-alanine dipeptide to a para-aminobenzyl (PAB) self-immolative spacer to enable efficient payload release under lysosomal conditions. The product is supplied as a solid reagent intended for use in conjugation workflows and analytical characterization.
  • Cleavable peptide linker for ADCs.
  • Enables enzymatic release of payload in lysosomes.
  • Contains valine-alanine dipeptide and PAB self-immolative spacer.
  • High purity suitable for research applications.
  • Solid form with storage at 4°C; protect from light.
  • Available in multiple package sizes, including 1 G.
Catalog No. 50-002-38044