Learn More
ATCC L Wnt-5A; Transfected; Mouse (Mus musculus)
Supplier:  ATCC CRL2814
Encompass
L Wnt-5A is a cell line exhibiting fibroblast-like morphology that was isolated from the areolar adipose tissue of a mouse. They are presently the best source for the production of Wnt-5A conditioned medium. Since the conditioned medium contains other factors besides the Wnt-5A protein, it is necessary to control any experiments involving the Wnt-5A conditioned medium with the control conditioned medium from the parental cell line (ATCC CRL-2648). This cell line was deposited by NCRR Contract.
Catalog No. 50-238-2147