Learn More
Medchemexpress LLC L-Xylose-2-13C | 478506-63-7 | 99.0% | C413CH10O5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY78139S11MG
Encompass_Preferred
L-Xylose-2-13C is the 13C labeled L-Xylose. L-Xylose is a rare sugar and the levorotatory form of Xylose. This compound serves as a raw material for the synthesis of various drugs and bioactive molecules. It is intended for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
Catalog No. 50-004-35074