Learn More
Medchemexpress LLC HY-100671 10mg Medchemexpress, L002 CAS:321695-57-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10067110MG
Encompass_Preferred
Medchemexpress, HY-100671 10mg L002 CAS:321695-57-2 L002 is a potent, cell permeable, reversible and specific acetyltransferase p300 (KAT3B) inhibitor with an IC50 of 1.98 uM. L002 binds the acetyl-CoA pocket and competitively inhibits the FATp300 catalytic domain, blocks histone acetylation and p53 acetylation, and inhibits STAT3 activation. L002 has the potential for hypertension‐induced cardiac hypertrophy and fibrogenesis treatment[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7762