Learn More
Waters Corp L1 UPLC Columns Method Development Kit 2.1 mm x 50 mm, 4/pk
Supplier:  Waters Corp 176001605
Encompass_Preferred
The L1 UPLC method development kit combines 1.7 µm BEH C18, Shield RP18, and 1.8 µm HSS C18 and T3 column chemistries to accomodate your method development approach, enabling methods to be developed efficiently and effectively. By utilizing UPLC Technology for method development, a substantial improvement in throughput can be realized. This, in turn, reduces cost per sample and time of analysis considerably while maintaining or improving separation integrity, enabling products to be brought to market faster and improving the overall profitability of the assay.
Catalog No. 50-791-019
Add to Cart
Request a Quote