Learn More
Waters Corp Waters Corporation L1 UPLC Columns Kit, 3 mm x 50 mm, 1.7/1.8 μm, 4-pack
Supplier:  Waters Corp 176001885
Encompass_Preferred
The Waters L1 UPLC Columns Kit is designed for method development, offering columns with varied silanol activity and hydrophobicity within the US Pharmacopeia L1 classification. This kit simplifies chromatographic method development by providing reliable and robust separation results.
  • Includes BEH C18, BEH Shield RP18, HSS C18, and HSS T3 chemistries.
  • Columns feature particle sizes of 1.7 μm for BEH and 1.8 μm for HSS.
  • Each column measures 3.0 mm x 50 mm.
  • Kit contains 4 columns per pack.
  • Columns offer varied silanol activity and hydrophobicity.
  • Designed to facilitate method development for L1 classification.
Catalog No. 50-791-062
Add to Cart
Request a Quote