Learn More
ATCC L1210; Leukemia; Mouse (Mus musculus)
Supplier:  ATCC CCL219
Encompass
L1210 is a cell line exhibiting lymphoblast morphology that was isolated from the skin of an eight-month-old, female mouse with lymphocytic leukemia, developed after skin paintings with 0.2% methylcholanthrene in ether. This cell line was deposited by EM Jensen and can be used in immune system disorder research.
Catalog No. 50-238-5084