Learn More
Carbosynth LLC. L17E Cytosolic Delivery Peptide (0.5 mg vial), 1ea, MW: 2859.4, Molecular formula: C134H220N38O31, CAS# 1898254-09-5
SDP
Supplier:  Carbosynth LLC. PLT3409V0.5MG
Encompass_Preferred
Synonyms: L17E, L17, Ile-Trp-Leu-Thr-Ala-Leu-Lys-Phe-Leu-Gly-Lys-His-Ala-Ala-Lys-His-Glu-Ala-Lys-Gln-Gln-Leu-Ser-Lys-Leu-NH2, Purity: , ≥96.0% (HPLC), Application: Peptide for Transport to Cytoplasma, Storage Temperature: -20 °C, References: M. Akishiba, et al., Nature Chemistry, 9, 751 (2017). (Original)
Catalog No. 50-168-6633