Learn More
TARGETMOL CHEMICALS INC L189 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T451725MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. L189 is a human DNA ligase inhibitor, inhibits hLigI/III/IV (IC50 5/9/5 uM). Purity 99.72%
Catalog No. 50-288-0189