Learn More
Medchemexpress LLC L5-DA | 2986876-41-7 | 99.0% | C32H34N6O2 | 10MG

Supplier: Medchemexpress LLC HY14471210MG
L5-DA is a small-molecule G-quadruplex stabilizer used in biochemical and cellular research to stabilize G4 DNA structures and probe G4-related biology. It shows cellular activity in model assays and is supplied with high chemical purity; researchers should follow supplier guidance for solubility and storage when preparing in vitro or in vivo formulations.
- Stabilizes G-quadruplex DNA structures.
- Reported cytotoxic activity in HeLa cells (IC50 4.3 μM).
- High purity (99.0%).
- Soluble in DMSO (≈25 mg/mL) for in vitro use.
- Formulation guidance provided for in vivo suspended solutions.
- Store sealed at 4°C and protect from moisture.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.