Learn More
Selleck Chemical LLC L755507 5mg 159182-43-1
SDP
Supplier:  Selleck Chemical LLC S79745MG
Encompass_Preferred
L-755,507 is characterized as a potent and selective ?3 adrenergic receptor partial agonist with EC50 of 0.43 nM. It is also recently identified to enhance CRISPR-mediated homology-directed repair (HDR) efficiency in human induced pluripotent stem cells (iPSCs) and other cell types. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5494