Learn More
Medchemexpress LLC L803-mts (Myristoylated L 803) | 706785-93-5 | 98.7% | 1465.63 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP11735MG
Encompass_Preferred
L803-mts (Myristoylated L 803) is a selective and substrate-competitive GSK-3 peptide inhibitor (IC50: 40 μM). It reduces Aβ deposits and ameliorates cognitive deficits in 5XFAD mice. L803-mts also shows an antidepressive effect in the forced swimming test.
  • Selective and substrate-competitive GSK-3 peptide inhibitor (IC50: 40 μM)
  • Reduces Aβ deposits
  • Ameliorates cognitive deficits in 5XFAD mice
  • Shows antidepressive effect in the forced swimming test
Catalog No. 50-004-22163