Learn More
Ace Glass, Inc. Lab bottle, Duran, 500mL, amber, GL 45, no ring or cap, cs/10
SDP
Supplier:  Ace Glass, Inc. 5539258
Encompass
Chemical-resistant and stable 500mL Duran laboratory bottles, fabricated from borosilicate glass and ambered. Supplied graduated with GL45 thread. Does not include cap or pouring ring. Can be hot-air sterilized or autoclaved. Case/10.
Catalog No. 50-978-390