Learn More
Research Products International Corp Laboratory Grade Rubber Stoppers w/Hole, BLK SBR Rubber, Size #13, 68 x 58 x 25mm, 4 per Package
SDP
Supplier:  Research Products International Corp 259150H13
Encompass
Laboratory Grade Rubber Stoppers with hole are offered in a wide range of common sizes for most laboratory applications. Available standard black SBR rubber. Material can typically withstand temperatures up to about 100°C (212°F). Rubber stoppers with hole are molded in the USA according to standard industry size specification. 
Packaged in approximately one pound packages.
Catalog No. 50-222-9705