Learn More
GA International Cryogenic Multi Color Labels for 1.5 mL Microtubes 1.02" x 0.4" / 25.9mm x 10.2mm Rectangle White From -196°C to +110°C (-321°F to +230°F)
Supplier:  GA International JTA2610WH
Encompass_Preferred
Ideal for the identification of vials, microtubes, plastic bags, and other containers used in deep-freeze storage and color-coding in harsh laboratory environments. These labels are designed for reliable performance in extreme cold conditions, including liquid nitrogen exposure.They write easily with permanent markers—recommended options include MP-1, F-1, and SM-1.
Catalog No. NC0495152