Learn More
GA International Cryogenic Labels for Laser Printers 0.43" / 10.9mm Circle Lavender From -80°C to +120°C (-112°F to +248°F)
Supplier:  GA International CLA48LA
Encompass_Preferred
Coated with a unique cryo adhesive, Cryo-LazrTAG™ thermoplastic waterproof labels are specifically designed to permanently adhere to the caps of 1.5 and 2ml vials and microtubes that will undergo long-term deep-freeze storage. These waterproof, durable labels are supplied in US letter format, allowing for easy, jam-free printing with standard desktop laser printers. Resistant to wear, handling, and mild laboratory chemicals, including swabbing with alcohols (isopropanol, ethanol) and surface sanitizing wipes, they deliver consistent reliability in demanding lab settings.*Given the small diameter of these dot labels, they are not recommended for liquid nitrogen storage. For tube cap labelling, we recommend thermal transfer or writable NitroTAG.
Catalog No. 50-005-71079